Assay Details
| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511698.2 | XP_003511746.1 | 2 - 3 | 486 | 65 | |
| GenBank mRNA | JI890569.1 | - | 2 - 3 | 486 | 65 | |
| JP062444.1 | - | 2 - 3 | 486 | 65 | ||
More Information
Set Membership: |
|

Mesdc2
Genomic Map