Assay Details
| Gene Symbol: Mpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650704.1 | XP_007648894.1 | 3 - 4 | 348 | 64 | |
| XM_007650705.1 | XP_007648895.1 | 3 - 4 | 348 | 64 | ||
| XM_007650706.1 | XP_007648896.1 | 3 - 4 | 334 | 64 | ||
| GenBank mRNA | JI889922.1 | - | 441 | 64 | ||
More Information
Set Membership: |
|

Mpc1
Genomic Map