Assay Details
| Gene Symbol: Cd300e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495708.1 | XP_003495756.1 | 2 - 3 | 388 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd300e| Gene Symbol: Cd300e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495708.1 | XP_003495756.1 | 2 - 3 | 388 | 85 | |
Set Membership: |
|