Assay Details
| Gene Symbol: Avpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497760.2 | XP_003497808.1 | 2 - 3 | 440 | 61 | |
| GenBank mRNA | JI869621.1 | - | 2 - 3 | 442 | 61 | |
| JP040180.1 | - | 2 - 3 | 442 | 61 | ||
More Information
Set Membership: |
|

Avpi1
Genomic Map