Assay Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506837.2 | XP_003506885.2 | 2 - 3 | 157 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ten1| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506837.2 | XP_003506885.2 | 2 - 3 | 157 | 78 | |