Assay Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612897.1 | XP_007611087.1 | 3 - 4 | 1028 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdyl| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612897.1 | XP_007611087.1 | 3 - 4 | 1028 | 59 | |
Set Membership: |
|