Assay Details
| Gene Symbol: LOC100766510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643956.1 | XP_007642146.1 | 7 - 8 | 808 | 84 | |
| GenBank mRNA | JI887349.1 | - | 7 - 8 | 712 | 84 | |
| JP060375.1 | - | 7 - 8 | 712 | 84 | ||
More Information
Set Membership: |
|

LOC100766510
Genomic Map