Assay Details
| Gene Symbol: LOC100766510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643956.1 | XP_007642146.1 | 16 - 17 | 6093 | 95 | |
| GenBank mRNA | JI879824.1 | - | 3 - 4 | 333 | 95 | |
More Information
Set Membership: |
|

LOC100766510
Genomic Map