Assay Details
| Gene Symbol: LOC100766510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI887349.1 | - | 5 - 6 | 517 | 89 | |
| JP060375.1 | - | 5 - 6 | 517 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100766510| Gene Symbol: LOC100766510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI887349.1 | - | 5 - 6 | 517 | 89 | |
| JP060375.1 | - | 5 - 6 | 517 | 89 | ||