Assay Details
| Gene Symbol: LOC100766207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640524.1 | XP_007638714.1 | 8 - 9 | 809 | 87 | |
| GenBank mRNA | JI891559.1 | - | 2 - 3 | 159 | 87 | |
| JP044489.1 | - | 2 - 3 | 159 | 87 | ||
More Information
Set Membership: |
|

LOC100766207
Genomic Map