Assay Details
| Gene Symbol: LOC100754898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646200.1 | XP_007644390.1 | 2 - 3 | 263 | 122 | |
| XM_007646201.1 | XP_007644391.1 | 2 - 3 | 263 | 122 | ||
| GenBank mRNA | JI889978.1 | - | 3 - 4 | 409 | 122 | |
| JP042359.1 | - | 3 - 4 | 409 | 122 | ||
More Information
Set Membership: |
|

LOC100754898
Genomic Map