Assay Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497807.2 | XP_003497855.1 | 6 - 7 | 1007 | 65 | |
| XM_007618040.1 | XP_007616230.1 | 7 - 8 | 743 | 65 | ||
| GenBank mRNA | JI896562.1 | - | 1 - 2 | 257 | 65 | |
More Information
Set Membership: |
|

Kcnip3
Genomic Map