Assay Details
| Gene Symbol: G6pc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504490.2 | XP_003504538.1 | 2 - 3 | 438 | 63 | |
| GenBank mRNA | JI866309.1 | - | 2 - 3 | 438 | 63 | |
| JP063229.1 | - | 2 - 3 | 438 | 63 | ||
More Information
Set Membership: |
|

G6pc3
Genomic Map