Assay Details
| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510639.1 | XP_003510687.1 | 2 - 3 | 1245 | 80 | |
| GenBank mRNA | JI871441.1 | - | 2 - 3 | 947 | 80 | |
| JP038213.1 | - | 2 - 3 | 947 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mepce| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510639.1 | XP_003510687.1 | 2 - 3 | 1245 | 80 | |
| GenBank mRNA | JI871441.1 | - | 2 - 3 | 947 | 80 | |
| JP038213.1 | - | 2 - 3 | 947 | 80 | ||