Assay Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512595.2 | XP_003512643.1 | 5 - 6 | 582 | 62 | |
| GenBank mRNA | JP063076.1 | - | 5 - 6 | 488 | 62 | |
More Information
Set Membership: |
|

Knstrn
Genomic Map