Assay Details
| Gene Symbol: LOC103158710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607744.1 | XP_007605934.1 | 5 - 6 | 674 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103158710| Gene Symbol: LOC103158710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607744.1 | XP_007605934.1 | 5 - 6 | 674 | 87 | |
Set Membership: |
|