Assay Details
| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649689.1 | XP_007647879.1 | 5 - 6 | 680 | 67 | |
| GenBank mRNA | JI893025.1 | - | 5 - 6 | 732 | 67 | |
More Information
Set Membership: |
|

Mrpl47
Genomic Map