Assay Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652148.1 | XP_007650338.1 | 3 - 4 | 324 | 73 | |
| GenBank mRNA | JP062055.1 | - | 2 - 3 | 214 | 73 | |
More Information
Set Membership: |
|

Nek6
Genomic Map