Assay Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647149.1 | XP_007645339.1 | 1 - 2 | 352 | 91 | |
| GenBank mRNA | JI862317.1 | - | 1 - 2 | 352 | 91 | |
More Information
Set Membership: |
|

Rnf138
Genomic Map