Assay Details
| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644676.1 | XP_007642866.1 | 6 - 7 | 1363 | 67 | |
| GenBank mRNA | JP064792.1 | - | 2 - 3 | 223 | 67 | |
More Information
Set Membership: |
|

Rnf8
Genomic Map