Assay Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495304.2 | XP_003495352.1 | 6 - 7 | 949 | 62 | |
| XM_007634958.1 | XP_007633148.1 | 6 - 7 | 937 | 62 | ||
| GenBank mRNA | JP063899.1 | - | 6 - 7 | 831 | 62 | |
More Information
Set Membership: |
|

Oxnad1
Genomic Map