Assay Details
| Gene Symbol: Gpn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498428.2 | XP_003498476.1 | 3 - 4 | 382 | 72 | |
| XM_007627854.1 | XP_007626044.1 | 3 - 4 | 312 | 72 | ||
| GenBank mRNA | JI879088.1 | - | 3 - 4 | 370 | 72 | |
| JP038075.1 | - | 3 - 4 | 370 | 72 | ||
More Information
Set Membership: |
|

Gpn3
Genomic Map