Assay Details
| Gene Symbol: Znf438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505021.2 | XP_003505069.1 | 6 - 7 | 2338 | 104 | |
| XM_007645512.1 | XP_007643702.1 | 7 - 8 | 2373 | 104 | ||
| XM_007645513.1 | XP_007643703.1 | 7 - 8 | 2315 | 104 | ||
| XM_007645514.1 | XP_007643704.1 | 7 - 8 | 2576 | 104 | ||
| XM_007645515.1 | XP_007643705.1 | 8 - 9 | 2490 | 104 | ||
| XM_007645516.1 | XP_007643706.1 | 7 - 8 | 2455 | 104 | ||
| XM_007645517.1 | XP_007643707.1 | 6 - 7 | 2286 | 104 | ||
| XM_007645518.1 | XP_007643708.1 | 7 - 8 | 2403 | 104 | ||
| XM_007645519.1 | XP_007643709.1 | 3 - 4 | 2046 | 104 | ||
| XM_007645520.1 | XP_007643710.1 | 7 - 8 | 2375 | 104 | ||
More Information
Set Membership: |
|

Znf438
Genomic Map