Assay Details
| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508737.2 | XP_003508785.1 | 2 - 3 | 1035 | 67 | |
| GenBank mRNA | JI875444.1 | - | 1 - 2 | 51 | 67 | |
| JP046005.1 | - | 1 - 2 | 51 | 67 | ||
More Information
Set Membership: |
|

Rom1
Genomic Map