Assay Details
| Gene Symbol: Smndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512590.2 | XP_003512638.1 | 4 - 5 | 598 | 64 | |
| XM_007653049.1 | XP_007651239.1 | 5 - 6 | 989 | 64 | ||
| GenBank mRNA | JI868692.1 | - | 4 - 5 | 581 | 64 | |
| JP050677.1 | - | 3 - 4 | 460 | 64 | ||
More Information
Set Membership: |
|

Smndc1
Genomic Map