Assay Details
Gene Symbol: LOC100754535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648438.1 | XP_007646628.1 | 4 - 5 | 491 | 75 | |
XM_007648439.1 | XP_007646629.1 | 5 - 6 | 437 | 75 | ||
XM_007648440.1 | XP_007646630.1 | 5 - 6 | 521 | 75 | ||
XM_007648441.1 | XP_007646631.1 | 4 - 5 | 245 | 75 | ||
XM_007648442.1 | XP_007646632.1 | 5 - 6 | 369 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
