Assay Details
| Gene Symbol: LOC100754535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648438.1 | XP_007646628.1 | 5 - 6 | 631 | 72 | |
| XM_007648439.1 | XP_007646629.1 | 6 - 7 | 577 | 72 | ||
| XM_007648440.1 | XP_007646630.1 | 6 - 7 | 661 | 72 | ||
| XM_007648441.1 | XP_007646631.1 | 5 - 6 | 385 | 72 | ||
| XM_007648442.1 | XP_007646632.1 | 6 - 7 | 509 | 72 | ||
More Information
Set Membership: |
|

LOC100754535
Genomic Map