Assay Details
| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640674.1 | XP_007638864.1 | 5 - 6 | 5919 | 74 | |
| XM_007640675.1 | XP_007638865.1 | 5 - 6 | 5882 | 74 | ||
| XM_007640676.1 | XP_007638866.1 | 5 - 6 | 5915 | 74 | ||
| GenBank mRNA | JI886563.1 | - | 2 - 3 | 342 | 74 | |
| JP039795.1 | - | 2 - 3 | 342 | 74 | ||
More Information
Set Membership: |
|

Hivep2
Genomic Map