Assay Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508145.2 | XP_003508193.1 | 5 - 6 | 1031 | 59 | |
| GenBank mRNA | JI880256.1 | - | 5 - 6 | 673 | 59 | |
| JP042986.1 | - | 5 - 6 | 673 | 59 | ||
More Information
Set Membership: |
|

Wwtr1
Genomic Map