Assay Details
| Gene Symbol: LOC100759096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513941.1 | XP_003513989.1 | 1 - 2 | 164 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759096| Gene Symbol: LOC100759096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513941.1 | XP_003513989.1 | 1 - 2 | 164 | 96 | |
Set Membership: |
|