Assay Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650044.1 | XP_007648234.1 | 2 - 3 | 253 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st3| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650044.1 | XP_007648234.1 | 2 - 3 | 253 | 71 | |
Set Membership: |
|