Assay Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646546.1 | XP_007644736.1 | 26 - 27 | 2654 | 67 | |
| GenBank mRNA | JI866061.1 | - | 14 - 15 | 1396 | 67 | |
More Information
Set Membership: |
|

Rb1
Genomic Map