Assay Details
| Gene Symbol: Runx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645808.1 | XP_007643998.1 | 4 - 5 | 946 | 58 | |
| GenBank mRNA | JI896450.1 | - | 2 - 3 | 263 | 58 | |
More Information
Set Membership: |
|

Runx1
Genomic Map