Assay Details
| Gene Symbol: Faxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643432.1 | XP_007641622.1 | 4 - 5 | 374 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faxdc2| Gene Symbol: Faxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643432.1 | XP_007641622.1 | 4 - 5 | 374 | 53 | |
Set Membership: |
|