Assay Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645257.1 | XP_007643447.1 | 3 - 4 | 206 | 70 | |
| GenBank mRNA | JI878700.1 | - | 5 - 6 | 296 | 70 | |
More Information
Set Membership: |
|

Urgcp
Genomic Map