Assay Details
| Gene Symbol: Chkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515462.2 | XP_003515510.1 | 4 - 5 | 746 | 62 | |
| XM_007607632.1 | XP_007605822.1 | 4 - 5 | 437 | 62 | ||
| GenBank mRNA | JI864817.1 | - | 1 - 2 | 161 | 62 | |
More Information
Set Membership: |
|

Chkb
Genomic Map