Assay Details
| Gene Symbol: LOC100762908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642281.1 | XP_007640471.1 | 1 | 701 | 68 | |
| GenBank mRNA | JP045226.1 | - | 4 - 5 | 405 | 68 | |
More Information
Set Membership: |
|

LOC100762908
Genomic Map