Assay Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645795.1 | XP_007643985.1 | 4 - 5 | 702 | 74 | |
| GenBank mRNA | JI880565.1 | - | 4 - 5 | 806 | 74 | |
More Information
Set Membership: |
|

Tbp
Genomic Map