Assay Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495087.2 | XP_003495135.1 | 11 - 12 | 1589 | 62 | |
| XM_007652446.1 | XP_007650636.1 | 9 - 10 | 1409 | 62 | ||
| XM_007652488.1 | XP_007650678.1 | 9 - 10 | 1427 | 62 | ||
More Information
Set Membership: |
|

Cbfa2t3
Genomic Map