Assay Details
| Gene Symbol: Erp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498421.2 | XP_003498469.1 | 1 - 2 | 236 | 67 | |
| GenBank mRNA | JI891681.1 | - | 1 - 2 | 198 | 67 | |
| JP040018.1 | - | 1 - 2 | 198 | 67 | ||
More Information
Set Membership: |
|

Erp29
Genomic Map