Assay Details
| Gene Symbol: Rd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498573.1 | XP_003498621.1 | 1 - 2 | 338 | 116 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  Probe spans exons | 
 Back To Top
 Back To Top
 Rd3
Rd3 Genomic Map
Genomic Map Back To Top
 Back To Top