Assay Details
Gene Symbol: LOC100764246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500358.2 | XP_003500406.1 | 1 - 2 | 1048 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100764246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500358.2 | XP_003500406.1 | 1 - 2 | 1048 | 98 |