Assay Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506449.2 | XP_003506497.1 | 3 - 4 | 1104 | 57 | |
| XM_007646966.1 | XP_007645156.1 | 4 - 5 | 1003 | 57 | ||
| XM_007646967.1 | XP_007645157.1 | 4 - 5 | 1081 | 57 | ||
| XM_007646968.1 | XP_007645158.1 | 4 - 5 | 982 | 57 | ||
| XM_007646969.1 | XP_007645159.1 | 4 - 5 | 904 | 57 | ||
| XM_007646970.1 | XP_007645160.1 | 4 - 5 | 904 | 57 | ||
More Information
Set Membership: |
|

Apba3
Genomic Map