Assay Details
Gene Symbol: Rnf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508264.2 | XP_003508312.1 | 4 - 5 | 571 | 78 | |
XM_007648885.1 | XP_007647075.1 | 4 - 5 | 680 | 78 | ||
XM_007648886.1 | XP_007647076.1 | 4 - 5 | 609 | 78 | ||
XM_007648887.1 | XP_007647077.1 | 2 - 3 | 385 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
