Assay Details
| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648724.1 | XP_007646914.1 | 2 - 3 | 200 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ascl2| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648724.1 | XP_007646914.1 | 2 - 3 | 200 | 64 | |
Set Membership: |
|