Assay Details
| Gene Symbol: LOC100765785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655079.1 | XP_007653269.1 | 6 - 7 | 726 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765785| Gene Symbol: LOC100765785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655079.1 | XP_007653269.1 | 6 - 7 | 726 | 74 | |
Set Membership: |
|