Assay Details
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495392.2 | XP_003495440.1 | 3 - 4 | 446 | 60 | |
| GenBank mRNA | JI888615.1 | - | 2 - 3 | 323 | 60 | |
| JP042699.1 | - | 2 - 3 | 323 | 60 | ||
More Information
Set Membership: |
|

Pak2
Genomic Map