Assay Details
| Gene Symbol: G6pc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495933.1 | XP_003495981.1 | 2 - 3 | 325 | 70 | |
| XM_007645331.1 | XP_007643521.1 | 2 - 3 | 325 | 70 | ||
More Information
Set Membership: |
|

G6pc2
Genomic Map