Assay Details
| Gene Symbol: Znf445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500669.2 | XP_003500717.1 | 6 - 7 | 1171 | 81 | |
| XM_007641421.1 | XP_007639611.1 | 6 - 7 | 1258 | 81 | ||
| XM_007641422.1 | XP_007639612.1 | 7 - 8 | 1388 | 81 | ||
| XM_007641423.1 | XP_007639613.1 | 7 - 8 | 1301 | 81 | ||
| XM_007641424.1 | XP_007639614.1 | 6 - 7 | 1242 | 81 | ||
| XM_007641425.1 | XP_007639615.1 | 4 - 5 | 375 | 81 | ||
| GenBank mRNA | JI868013.1 | - | 3 - 4 | 563 | 81 | |
| JP051947.1 | - | 4 - 5 | 352 | 81 | ||
More Information
Set Membership: |
|

Znf445
Genomic Map