Assay Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500758.2 | XP_003500806.1 | 3 - 4 | 279 | 83 | |
| GenBank mRNA | JI877246.1 | - | 2 - 3 | 210 | 83 | |
| JP046167.1 | - | 3 - 4 | 352 | 83 | ||
More Information
Set Membership: |
|

Lsm6
Genomic Map